Learn More
HIV-1, gp120, Mouse anti-Viruses, Clone: 1G12, Abnova™
Mouse Monoclonal Antibody
Brand:  Abnova MAB0036-M08.50ug
Product Code. 16079715
  • 393.00€ / 50µg
Specifications
HIV-1, gp120
Monoclonal
Unconjugated
In PBS, pH 7.4
Recombinant Fc fusion protein corresponding to amino acids 255-477 of human Immunodeficiency Virus type 1 (HIV-1), CN54 gp120 outer domain.
RUO
Human
Antibody
IgG1 κ
ELISA, Western Blot
1G12
The optimal working dilution should be determined by the end user.
Mouse
50 μg
Primary
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Liquid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
HIV-1, gp120, Mouse anti-Viruses, Clone: 1G12, Abnova™ > 50μg; Unconjugated