missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA-DPA1 293T Cell Overexpression Lysate (Denatured), Abnova
Cell transient overexpression lysate for HLA-DPA1
Brand: Abnova H00003113-T01.100uL
This item is not returnable.
View return policy
Description
HLA-DPA1 belongs to the HLA class II alpha chain paralogues. This class II molecule is a heterodimer consisting of an alpha (DPA) and a beta (DPB) chain, both anchored in the membrane. It plays a central role in the immune system by presenting peptides derived from extracellular proteins.
- Major histocompatibility complex, class II, DP alpha 1
- Transfected cell line: 293T
- Plasmid: pCMV-HLA-DPA1 full-length
- Host: Human
- Theoretical molecular weight: 29.3kDa
- Storage buffer: 1x sample buffer (50mM Tris-HCl, 2% SDS, 10% glycerol, 300mM 2-mercaptoethanol, 0.01% bromophenol blue)
Western Blotting
Specifications
293T | |
HLA-DPA1 293T Cell Transient Overexpression Lysate(Denatured) | |
29.3 | |
pCMV-HLA-DPA1 full-length | |
100 μL | |
Human |
Western Blot | |
BC009956 | |
6.8 | |
Transient overexpression cell Lysate was tested with Anti-HLA-DPA1 antibody (H00003113-B01) by western blotting | |
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction