Learn More
Hoefer™ Blotting Paper
Cellulose papers precut for convenience
Brand: Hoefer™ TE46
This item is not returnable.
View return policy
Description
- For Western Blotting
Specifications
14.5 cm | |
TE42 or TE62 Tank Blotting Units | |
Cellulose |
21.5 cm | |
Sheet |
Product Content Correction
Product Title
Hoefer™ Blotting Paper > 14.5 x 21.5cm