Learn More
Hoefer™ Blotting Paper
Cellulose papers precut for convenience
Brand:  Hoefer™ TE76
 View more versions of this product
Product Code. 12069847
  • 74.65€ / Pack of 25
Estimated Shipment: 04-07-2025
Description
  • For Western Blotting
Specifications
Specifications
21 cm
TE77 Semi-Dry Blotting Units
Cellulose
25 cm
Sheet
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Hoefer™ Blotting Paper > 21 x 26cm