Learn More
Hoefer™ HE 99X Max Submarine Electrophoresis Unit
Offers no-mess gel casting without tape or dams
Brand: Hoefer™ HE99X
Includes:
Buffer chamber with electrodes and leveling feet, safety lid with leads, bubble level. Complete system (03-500-121) also includes 15 x 20cm gel casting tray and UV-transparent running tray; 1.5mm thick, 15-well comb; adjustable comb back that permits control of the depth of the sample wells
Description
- Unit accommodates a variety of gel lengths for maximum resolution of DNA fragments
- Agarose bridges are also accommodated, so gels can be run in nonsubmarine fashion
- Rugged design provides reliability and safety
Specifications
EN61010-1, UL61010A-1, CSA C22.2 1010.1, CE | |
Up to 80% | |
HE 99X Max Basic | |
113°F | |
40.5 oz. | |
14.1 x 7.1 x 5.5 in. (36 x 18.2 x 14 cm) | |
5.9, 3.9 or 7.8 in. | |
15, 10 or 20 cm | |
5.9 in. | |
15 cm | |
14 cm | |
20 w |
Horizontal Agarose Electrophoresis Unit | |
Buffer chamber assembly, safety lid with high voltage Leads, Foam gaskets, bubble level | |
60 | |
45°C | |
1200 mL | |
14.1 in. | |
36 cm | |
7.1 in. | |
18.2 cm | |
5.5 in. | |
200 V | |
100 mA |