missing translation for 'onlineSavingsMsg'
Learn More

Hoefer™ Maxi-Plus and Maxi-Plus Submarine Gel Electrophoresis Units

Ideal for high resolution analytical and preparative studies of nucleic acids

Brand:  Hoefer™ SUB25C

 View more versions of this product

Product Code. 12665786

  • 1230.00€ / Each
Estimated Shipment: 07-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Large format ideal for high resolution techniques
  • Multichannel pipette compatible combs
  • Color-coded and height adjustable combs
  • Buffer recirculation ports
  • Colored loading strips
  • UV-transparent running tray
  • Side handles for safe, easy transport
Specifications

Specifications

EN61010-1, CE
Up to 80%
624
45°C
3000 mL
22 in.
56 cm
13.7 in.
35 cm
3.5 in.
1000 V
500 mA
Submarine Gel Electrophoresis Unit
Buffer chamber assembly with cooling ports, safety lid with high voltage Leads, running tray with casting gates, 1mm thick, 16-well combs, colored loading strips, Buffer recirculation ports
113°F
101.4 oz.
22 x 13.7 x 3.5 in. (56 x 35 x 9 cm)
11.8 in.
30 cm
9.8 in.
25 cm
9 cm
500 w
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers