missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hoefer™ Maxi-Plus and Maxi-Plus Submarine Gel Electrophoresis Units
Ideal for high resolution analytical and preparative studies of nucleic acids
Brand: Hoefer™ SUB25
This item is not returnable.
View return policy
Description
- Large format ideal for high resolution techniques
- Multichannel pipette compatible combs
- Color-coded and height adjustable combs
- Buffer recirculation ports
- Colored loading strips
- UV-transparent running tray
- Side handles for safe, easy transport
Specifications
EN61010-1, CE | |
Maxi Plus Standard Submarine Gel Electrophoresis Unit | |
80% | |
624 | |
45°C | |
22.04 in. | |
56 cm | |
11.8 in. | |
1000 V | |
500 mA |
Nucleic Acid Analysis | |
25 x 30 cm | |
Buffer chamber assembly, safety lid with high voltage Leads, running tray with casting gasket, 1mm thick, 16-well combs, colored loading strips, Buffer recirculation ports | |
113°F | |
22.04 x 13.77 in. (56 x 35 cm) | |
9.8 in. | |
25 cm | |
30 cm | |
500 w |