Learn More
Sperian™ Honeywell™ A700 Series Clear Anti-Fog Safety Glasses
Click to view available options
:
Product Type: Safety Glasses; Lens Coating: FogBan
Description
- Minimizes lens fogging in extreme conditions
- Affordably priced and economical for your business
- Lightweight design with sport temples and soft temple tip pads
- Moulded universal nose bridge
- Provides a secure, wraparound fit for all-day comfort
- 8-base polycarbonate abrasion-resistant lenses
- Provides a 180° field of distortion-free vision
Specifications
Specifications
Lens Coating | Anti-fog |
Product Type | Safety Glasses |
Frame Color | Clear |
No. per Pack | 10/Pk. |
Lens Material | Polycarbonate |
Product Content Correction
Product Title
Sperian™ Honeywell™ A700 Series Clear Anti-Fog Safety Glasses >