Learn More
Sperian™ Honeywell™ Clearways Faceshield
Click to view available options
Product Type:
Browguard with Elastic Headband
Visor
Lens Coating:
Anti-fog
Uncoated
Lens Material:
Acetate
N/A
Polycarbonate
Description
- Recommended for the agriculture, chemical and forestry industries
- Protects against hazards including abrasion, airborne particles, biohazards, chemicals, impacts, splashes and more
- Safe, practical, economical integral visor fitting ensures extra protection
Order Info
Note: The image shows both the Browguard and the Visor. Both pieces must be ordered separately. See Browguard (11886980), and Visors (11806990, 12652733, or 11896980).
Specifications
Specifications
Lens Coating | Uncoated |
Product Type | Visor |
No. per Pack | 1 Ea. |
Lens Material | Acetate |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Documents
Scientific Resources
Brochures
Applications and solutions series: Safety edition
File size: 7.7 MB
Product Content Correction
Product Title
Sperian™ Honeywell™ Clearways Faceshield >