Learn More
Sperian™ Honeywell™ ARMAMAX AX™ Clear Lens Safety Goggles
Offering high levels of comfort and support. Sperian™ Honeywell™ ARMAMAX AX™ Clear Lens Safety Goggles fit comfortably over prescription eyewear.
11.00€ - 93.00€
Specifications
Certifications/Compliance | ISO 9001 / 2000, EN 166 1F |
---|---|
Lens Tint | Clear |
Product Type | Eye and Face Protection Spectacles |
For Use With (Application) | Over-the-glass spectacle, General use and protection against impact (45m/s), Mechanic work: Grinding, turning, milling |
Frame Color | Transparent |
Product Code | Brand | Lens Coating | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Lens Coating | Price | Quantity & Availability | |||||
11847221
|
Sperian™
1002221 |
Hardcoat | N/A | ||||||
12970405
|
Sperian™
1002221 |
Hardcoat |
11.00€
Each |
Estimated Shipment: 20-12-2024
Log in to see stock. |
|||||
11857221
|
Sperian™
1002224 |
Uncoated |
93.00€
Pack of 10 |
In Stock
Log in to see stock. |
|||||
Description
- Practical and durable
- Frosted side panels and browguard sections reduce glare/reflections
- Curved temples and a raised nose bridge offer a precise fit
- Available with or without hard lens coating
Specifications
ISO 9001 / 2000, EN 166 1F | |
Eye and Face Protection Spectacles | |
Transparent | |
Polycarbonate |
Clear | |
Over-the-glass spectacle, General use and protection against impact (45m/s), Mechanic work: Grinding, turning, milling | |
10/Pk. |