missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sperian™ Honeywell™ Viking Noise Blocking Ear Muffs
Click to view available options
Color:
Black, Red
Black, Silver
Description
- Alternative to cap-mounted earmuffs – wear with hard hats, face shields, respirators and other PPE
- Patented Air Flow Control™ technology delivers optimal attenuation across all frequencies without increasing earcup size or weight
- Inner-ventilated headband minimizes pressure on the head, breathes easier in warm/humid climates [V2 and V3 only]
- Non-deforming outer headband withstands rough treatment in the toughest workplaces
- Snap-in ear cushions make replacement quick and easy
Specifications
Specifications
Color | Black, Red |
Certifications/Compliance | PPE CAT II, ISO 9001/2000, 29 CRF 1910.95, 2003/10/EC, 89/686/EEC |
For Use With (Application) | Noise Blocking Ear Muff |
Product Type | Hearing Protection |
Dielectric | Yes |
Style | Multi-position |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'