missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Hormones: Hydrocortisone
Brand: MP Biomedicals™ 0219456901
View more versions of this product
This item is not returnable.
View return policy
Description
For research use only. Not intended for therapeutic use.
Specifications
15° to 30°C | |
Sterile | |
Soy beans |
Hydrocortisone | |
1 g |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Warning
Hazard Category
- Reproductive toxicity Category 2
Hazard Statement
- H361-Suspected of damaging fertility or the unborn child.
Precautionary Statement
- P501-Dispose of contents/container to in accordance with local/regional/national/international regulations.
- P103-Read label before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P102-Keep out of reach of children.
- P101-If medical advice is needed, have product container or label at hand.
- P405-Store locked up.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction