Learn More
Grant Instruments™ HOSE100 - General purpose tubing and insulation kit.
HOSE100 General purpose tubing and insulation kit -40 to 100°C (2 x 2 m). Includes clips and connector for Grant Instruments LT ecocool and Optima thermostats.
Brand: Grant Instruments™ HOSE100
This item is not returnable.
View return policy
Specifications
2 x 2 m general purpose insulated tubing -40°C to +100°C, assembled with LT ecocool/OptimaTM pump outlet plate and simple tube clips, no tools required. Can be cut to length. 10 mm ID, 14 mm OD. |