missing translation for 'onlineSavingsMsg'
Learn More

Gestigkeit Harry™ Hotplate with Four Heating Positions, Cast-iron

Shape: Round, Size: 3.35 in. dia.

Brand:  Gestigkeit Harry™ SGK4

Product Code. 10416511

  • 1335.00€ / Each
Estimated Shipment: 11-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Shape: Round; Size: 3.35 in. dia.
Specifications

Specifications

Hot Plate
Round
Cast Iron
155 mm
145 mm
4.8 kg
230 V, 50/60 Hz, 1800 w
4
230/240 V
3.35 in. dia.
10.6 lb.
485 mm
1800 W
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers