missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSP27 Antibody (rHSPB1/7298), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-24296AF405
This item is not returnable.
View return policy
Description
HSP27 Monoclonal antibody specifically detects HSP27 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
HSP27 | |
Monoclonal | |
Alexa Fluor 405 | |
28 kDa heat shock protein, DKFZp586P1322, Estrogen-regulated 24 kDa protein, Heat shock 27 kDa protein, heat shock 27kD protein 1, heat shock 27kDa protein 1, heat shock protein beta-1, HMN2B, HS.76067, Hsp25, HSP27HSP 27, HSP28CMT2F, HspB1, SRP27, Stress-responsive protein 27 | |
Recombinant human full-length HSP27 protein | |
0.1 mL | |
Cancer, Cell Biology, Cell Cycle and Replication, Golgi Apparatus Markers, Membrane Trafficking and Chaperones, Neuroscience, Phospho Specific | |
3315 | |
Store at 4°C in the dark. | |
IgG1 Îş |
Immunohistochemistry (Paraffin) | |
rHSPB1/7298 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
HSP27 Antibody (rHSPB1/7298), Alexa Fluor™ 405, Novus Biologicals™ > 0.1 mL, Alexa Fluor 405
Spot an opportunity for improvement?Share a Content Correction