Learn More
Thermo Scientific™ HTM Broth
Perform high quality susceptibility testing with a range of supplies, including the Thermo Scientific™ HTM Broth, to enhance your Thermo Scientific™ Sensititre™ test program.
Brand:  Thermo Scientific™ YT3470
67.91 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 16531840
  • 69.65€ / Pack of 2
Description
Perform high-quality susceptibility testing with a complete range of supplies, including the Thermo Scientific™ HTM Broth, to complement and enhance your Thermo Scientific™ Sensititre™ test program.
Not all products are available for sale in all territories. Please inquire.
VersaTREK™ and Sensititre products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
HTM Broth
Microbial Susceptibility
11 mL/Bottle
Microbiology Media
10
11 mL
SDS
Documents Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
4 results found
Lot Number Certificate Type Date Catalog Number
Lot Number264235 Certificate TypeCertificate of Analysis Date10/05/2025 Catalog NumberYT3470
Lot Number249590 Certificate TypeCertificate of Analysis Date31/03/2025 Catalog NumberYT3470
Lot Number241136 Certificate TypeCertificate of Analysis Date17/03/2025 Catalog NumberYT3470
Lot Number225061 Certificate TypeCertificate of Analysis Date14/02/2025 Catalog NumberYT3470
Product Title
Thermo Scientific™ HTM Broth >