Learn More
R&D Systems™ Human Apolipoprotein H/ApoH (NP_000033) VersaClone cDNA
Brand: R&D Systems™ RDC2887
This item is not returnable.
View return policy
Specifications
NP_000033 | |
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. | |
10 μg |
350 | |
Human |