missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human beta-1,3-Glucuronyltransferase 1/B3GAT1 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB85603-SP
This item is not returnable.
View return policy
Description
beta-1,3-Glucuronyltransferase 1/B3GAT1 Monoclonal antibody specifically detects beta-1,3-Glucuronyltransferase 1/B3GAT1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry
Specifications
beta-1,3-Glucuronyltransferase 1/B3GAT1 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 ÎĽm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 ÎĽm filtered solution in PBS. | |
Beta-1,3-glucuronyltransferase 1, beta-1,3-glucuronyltransferase 1 (glucuronosyltransferase P), CD57, EC 2.4.1.135, galactosylgalactosylxylosylprotein 3-beta-glucuronosyltransferase 1, GlcAT-P, GLCATP, GLCATPGLCUATP, glcUAT-P, GLCUATP, Glucuronosyltransferase P, HNK1, LEU7, NK1, NK-1, UDP-GlcUA:glycoprotein beta-1,3-glucuronyltransferase | |
Mouse myeloma cell line NS0-derived recombinant rat beta -1,3-Glucuronyltransferase 1/B3GAT1, Asp75-Ile347, Accession # NP_445455 | |
25 ÎĽg | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
IgG2b |
Immunohistochemistry, Immunocytochemistry | |
1056734 | |
Immunohistochemistry 5-25 ug/mL, Immunocytochemistry 8-25 ug/mL | |
NP_445455 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
27087 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Human beta-1,3-Glucuronyltransferase 1/B3GAT1 Antibody, R&D Systems™ > 25 μg; Unconjugated
Spot an opportunity for improvement?Share a Content Correction