missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Biomarker Premixed Magnetic Luminex™ Performance Assay Kit A
Earn double rewards for reviews
1360.00€ - 2490.00€
Specifications
Species | Human |
---|---|
Binding Target | Multiplex Cytokine Assay Kits |
For Use With (Application) | ELISA |
Content And Storage | Store the unopened product at 2 - 8 °C. Do not use past expiration date. |
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
18730749
|
R&D Systems™
FCSTM13-07 |
7 Plex |
2490.00€
7 sets |
Estimated Shipment: 28-03-2025
Log in to see stock. |
||||||
18779667
|
R&D Systems™
FCSTM13-05 |
5 Plex |
2040.00€
5 sets |
Estimated Shipment: 28-03-2025
Log in to see stock. |
||||||
18769558
|
R&D Systems™
FCSTM13-02 |
2 Plex |
1360.00€
2 sets |
Estimated Shipment: 28-03-2025
Log in to see stock. |
||||||
Description
Specifications
Human | |
Multiplex Cytokine Assay Kits | |
ELISA | |
Store the unopened product at 2 - 8 °C. Do not use past expiration date. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
R&D Systems™ Human Biomarker Premixed Magnetic Luminex™ Performance Assay Kit A