Learn More
Novus Biologicals™ Human Brain Amygdala Whole Tissue Lysate (Adult Whole Normal)
Brand: Novus Biologicals™ NB820-59178
This item is not returnable.
View return policy
Description
The Human Brain Amygdala Whole Tissue Lysate (Adult Whole Normal) is derived from human. It has been validated for the following applications: Western Blot, Immunoprecipitation.
Specifications
Western Blot | |
1 mg | |
Total Protein - Human Adult Normal Tissue: Brain: Amygdala | |
Normal | |
Human | |
Human Brain Amygdala Whole Tissue Lysate (Adult Normal) | |
Adult |
Nervous | |
HEPES (pH 7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, Sodium deoxycholate, NP-40, and a cocktail of protease inhibitors | |
Whole | |
Human | |
5 mg/mL | |
-80°C. Avoid freeze-thaw cycles. | |
Native |
Product Content Correction
Product Title
Novus Biologicals™ Human Brain Amygdala Whole Tissue Lysate (Adult Whole Normal) > 1mg