missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human CD112 (PVRL2) Fc Chimera, Recombinant Protein
HEK293 cells Recombinant Protein
Brand: Invitrogen™ A42496
This item is not returnable.
View return policy
Description
This gene encodes a single-pass type I membrane glycoprotein with two Ig-like C2-type domains and an Ig-like V-type domain. This protein is one of the plasma membrane components of adherens junctions. It also serves as an entry for certain mutant strains of herpes simplex virus and pseudorabies virus, and it is involved in cell to cell spreading of these viruses. Variations in this gene have been associated with differences in the severity of multiple sclerosis. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.Specifications
Q92692 | |
protein with 0.02% Tween 80, 140 mM NaCl, 2.7 mM KCl, 5% mannitol, 5% trehalose and no preservative; pH 7.4 | |
Human CD112 (PVRL2) Fc Chimera | |
HEK293 cells | |
RUO | |
AI325026; AI987993; Cd112; Herpes virus entry mediator B; herpesvirus entry mediator B; herpesvirus entry protein B; hveB; mHveB; Mph; Murine herpes virus entry protein B; murine herpesvirus entry protein B; nectin cell adhesion molecule 2; NECTIN2; Nectin-2; Poliovirus receptor homolog; poliovirus receptor related 2; poliovirus receptor-like 2; poliovirus receptor-related 2; poliovirus receptor-related 2 (herpesvirus entry mediator B); poliovirus receptor-related protein 2; poliovirus sensitivity; PRR2; Pvr; Pvrl2; PVRR2; Pvs | |
Human | |
Recombinant | |
>95% |
Control, ELISA, Functional Assay, Immunohistochemistry, Western Blot | |
5819 | |
5 x 50 μg | |
-20°C | |
<1 EU/μg | |
NECTIN2 | |
Unconjugated | |
Lyophilized |