Learn More
Invitrogen™ Human CD63 ELISA Kit
Human CD63 quantitates human CD63 in serum, plasma, supernatant.
Brand:  Invitrogen™ EH95RB
 View more versions of this product
Product Code. 16853020
  • 656.00€ / 96 tests
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Human CD63 quantitates human CD63 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human CD63.
CD63 (LAMP-3, lysosome-associated membrane protein-3), a glycoprotein of tetraspanin family, is present in late endosomes, lysosomes and secretory vesicles of various cell types. CD63 is also present in the plasma membrane, usually following cell activation. Hence, CD63 has become a widely used basophil activation marker. In mast cells, however, CD63 exposition does not need their activation. CD63 interacts with integrins and affects phagocytosis and cell migration, it is also involved in H/K-ATPase trafficking regulation of ROMK1 channels. CD63 also serves as a T-cell costimulation molecule. Expression of CD63 can be used for predicting the prognosis in earlier stages of carcinomas. CD63 is expressed on activated platelets, and is a lysosomal membrane glycoprotein that is translocated to plasma membrane after platelet activation. CD63 is also present in monocytes and macrophages and is weakly expressed on granulocytes, B, and T cells. CD63 is identical to the melanoma-associated antigen which is ME491 and to the platelet antigen PTLGP40. Diseases associated with CD63 dysfunction include melanoma and Hermansky-Pudlak Syndrome.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
C75951; CD 63; CD63; CD63 antigen; CD63 antigen (melanoma 1 antigen); Cd63 molecule; CD63 protein; granulophysin; LAMP-3; Lysosomal-associated membrane protein 3; lysosome-associated membrane glycoprotein 3; mast cell antigen AD1; ME491; melanoma 1 antigen; melanoma-associated antigen ME491; melanoma-associated antigen MLA1; MLA1; Ocular melanoma-associated antigen; OMA81H; tetraspanin-30; TSPAN30; Tspan-30
14 pg/mL
ELISA
Colorimetric Microplate Reader
CD63
<10%
HRP
Research Use Only
2°C to 8°C
1 hr 20 min
14 to 3,000 pg/mL
Biotin
Plasma, Serum, Supernatant
P08962
<12%
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
96 Tests
Serum, 50 μL; Plasma, 50 μL; Supernatant, 50 μL
CD63
4 hr 45 min