Learn More
Invitrogen™ Human CEA ProQuantum Immunoassay Kit
The ProQuantum Human CEA Immunoassay Kit is designed to provide quantitative measurements of human CEA in small sample volumes.
764.00€
Specifications
Accession Number | 1600029H12Rik; Carcinoembryonic antigen; carcinoembryonic antigen related cell adhesion molecule 5; carcinoembryonic antigen-related cell adhesion molecule 5; CD66e; CEA; CEACAM5; meconium antigen 100; OTTHUMP00000199033; Pregnancy-specific glycoprotein 30; Psg30; sCD66e; soluble CD66e |
---|---|
Assay Range | 0.32 to 25,000 pg/mL |
Assay Sensitivity | <0.3 pg/mL |
Product Type | ELISA |
For Use With (Equipment) | qPCR Instruments |
Description
Utilizing proximity-based amplification technology, the assay combines the analyte specificity of high-affinity antibody-antigen binding with the signal detection and amplification capabilities of real-time PCR to achieve a simple yet powerful next-generation protein quantitation platform. A user-friendly workflow combined with intuitive software for analytics enables sample-to-answer in just 2 hours.
- High sensitivity-detect low levels of protein with greater sensitivity than traditional methods
- Broad dynamic range-5 logarithmic units, minimizing sample dilutions to ensure they fall within the range
- Small sample consumption-use 2–5 μL of sample (compared to 75 μL for triplicate wells with other methods)
- Fast, easy workflow-2 hours from sample to answer, with no wash steps
- No proprietary instrument to purchase-runs on any real-time PCR instrument
Specifications
1600029H12Rik; Carcinoembryonic antigen; carcinoembryonic antigen related cell adhesion molecule 5; carcinoembryonic antigen-related cell adhesion molecule 5; CD66e; CEA; CEACAM5; meconium antigen 100; OTTHUMP00000199033; Pregnancy-specific glycoprotein 30; Psg30; sCD66e; soluble CD66e | |
<0.3 pg/mL | |
qPCR Instruments | |
Ceacam5 | |
Research Use Only | |
CD66e (CEACAM5) | |
2 hr |
0.32 to 25,000 pg/mL | |
ELISA | |
P06731 | |
Assay Dilution Buffer Antibody-Conjugate A Antibody-Conjugate B Antibody-Conjugate Dilution Buffer Standard Master Mix Ligase | |
-20°C | |
30 min |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.