Learn More
R&D Systems™ Human Contactin-6 (NP_055276) VersaClone cDNA
Contactin6
Brand: R&D Systems™ RDC1837
This item is not returnable.
View return policy
Description
The Human Contactin-6 (NP_055276) VersaClone cDNA clone contains the complete ORF for Contactin-6, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The Contactin-6 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.Order Info
Ships at ambient temperature.
Specifications
cDNA | |
NP_055276 | |
cDNA is provided in 10mM Tris-Cl, pH 8.5 | |
27255 | |
Human |
hContactin-6 | |
10 μg at 0.2 μg/μL | |
3100bp | |
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. | |
10 μg |
Product Content Correction
Product Title
R&D Systems™ Human Contactin-6 (NP_055276) VersaClone cDNA > Quantity: 10μg
For Research Use Only. Not for use in Humans.