Learn More
R&D Systems™ Human Cystatin C Magnetic Luminex™ Performance Assay
For use with the magnetic Kidney Biomarker Panel
Brand:  R&D Systems™ LHK1196
Product Code. 18471800
  • 227.00€ / 1 set
Explore more special offers
Description
Description
- Magnetic format allows for easier wash steps - Undergoes similar validation testing as Quantikine™ ELISA Kits - Precise and Specific - Requires a small sample volume (<50μL) - Run your assay in just 3-5 hours - Mass-calibrated standards for consistent results with every new lot of material - Enough reagents for one 96-well plate
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Magnetic Bead-based Multiplex Assay for the Luminex™ MAGPIX™ Platform
Human
Magnetic
23.00 - 16500pg/mL
CST3
Serum (10μL),EDTA Plasma (10μL),Heparin Plasma (10μL),Urine (10μL)
Store the unopened product at 2° to 8°C. Do not use past expiration date.
1 Kit
Cystatin C
57.1pg/mL
1471
Serum, Plasma (EDTA, Heparin), Urine
  • Microparticle Concentrate
  • Biotin-Antibody Concentrate
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
R&D Systems™ Human Cystatin C Magnetic Luminex™ Performance Assay > 1 Kit