Learn More
R&D Systems™ Human Epoxide Hydrolase 2/EPHX2 (NP_001970) VersaClone cDNA
R&D Systems™ Human Epoxide Hydrolase 2/EPHX2 (NP_001970) VersaClone cDNA is a member of the epoxide hydrolase family.
Brand:  R&D Systems™ RDC2252
Product Code. 18798888
  • 322.00€ / 10µg
Estimated Shipment: 10-12-2024
Description
The Human Epoxide Hydrolase 2/EPHX2 (NP_001970) VersaClone cDNA clone contains the complete ORF for EPHX2, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The EPHX2 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Specifications
Specifications
hEPHX2
10 μg at 0.2 μg/μL
1681bp
2053
Human
NP_001970
cDNA is provided in 10 mM Tris-Cl
8.5
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.