Learn More
Novus Biologicals™ Human FDA Standard Tissue MicroArray (Normal)
The panel contains 5 to 8 different tissues on one slide.
Brand:  Novus Biologicals™ NBP2-78123
Product Code. 18067053
  • 1060.00€ / Pack of 5
Estimated Shipment: 15-01-2025
Specifications
5 Slides
Human
Please bake slides at 60C for 30 minutes before use.