missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human GM-CSF High Sensitivity Magnetic Luminex™ Performance Assay
Luminex bead-based multiplex assays are designed to provide accurate, reproducible results for every target analyte.
Brand: R&D Systems™ LHSCM215
This item is not returnable.
View return policy
Description
- Magnetic format allows for easier wash steps - Undergoes similar validation testing as Quantikine™ ELISA Kits - Precise and Specific - Requires a small sample volume (<50μL) - Run your assay in just 3-5 hours - Mass-calibrated standards for consistent results with every new lot of material - Enough reagents for one 96-well plate
Specifications
Magnetic Bead-based Multiplex Assay for the Luminex™ MAGPIX™ Platform | |
Human | |
Magnetic | |
0.39 - 1600pg/mL | |
CSF2 | |
Serum (50μL),EDTA Plasma (50μL),Heparin Plasma (50μL) | |
|
1 Kit | |
GM-CSF | |
0.13pg/mL | |
1437 | |
Serum, Plasma (EDTA, Heparin) | |
colony stimulating factor 2 (granulocyte-macrophage), Colony-stimulating factor, CSF, CSF2, CSF-2, GMCSF, GM-CSF, GMCSFgranulocyte-macrophage colony-stimulating factor, granulocyte-macrophage colony stimulating factor, MGC131935, MGC138897, Molgramostim, molgramostin, sargramostim | |
Store the unopened product at 2° to 8°C. Do not use past expiration date. |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 6
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
R&D Systems™ Human GM-CSF High Sensitivity Magnetic Luminex™ Performance Assay > 1 Kit
Spot an opportunity for improvement?Share a Content Correction