Learn More
R&D Systems™ Human gp130 Magnetic Luminex™ Performance Assay
Brand:  R&D Systems™ LBAM228
Product Code. 18726749
  • 227.00€ / Each
Explore more special offers
Specifications
Magnetic Bead-based Multiplex Assay for the Luminex™ MAGPIX™ Platform
Human
3572
1 Kit
gp130/CD130
Store the unopened product at 2 - 8 °C. Do not use past expiration date.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
R&D Systems™ Human gp130 Magnetic Luminex™ Performance Assay > 1 Kit