Learn More
R&D Systems™ Human IFN-alpha/beta R1 (AAA52730) VersaClone cDNA
Contains gene inserts that are fully sequenced and produced with high quality
Brand:  R&D Systems™ RDC1352
Product Code. 18724958
  • 322.00€ / 10µg
Estimated Shipment: 14-11-2024
Description
This shuttle vector contains the complete ORF for the gene of interest, along with a Kozak consensus sequence for optimal translation initiation. It is inserted NotI to AscI. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector.
The Human IFN-alpha/beta R1 (AAA52730) VersaClone cDNA clone contains the complete ORF for IFN-alpha/beta R1, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The IFN-alpha/beta R1 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Order Info
Ships at ambient temperature
Specifications
Specifications
IFN-alpha/beta R1
10 μg at 0.2 μg/μL
1687bp
alpha-type antiviral protein, AVP, beta-type antiviral protein, CRF2-1, Cytokine receptor class-II member 1, Cytokine receptor family 2 member 1, human interferon-alpha receptor (HuIFN-alpha-Rec)10IFRC, IFN-alpha/beta receptor 1, IFN-alpha-REC, IFNAR, IFN-aR1, IFNAR1, IFNBR, IFNbR1, IFN-bR1, IFN-R-1, interferon (alpha, beta and omega) receptor 1, interferon alpha/beta receptor 1, interferon-alpha/beta receptor alpha chain, interferon-beta receptor 1, Type I interferon receptor 1
Human
AAA52730
cDNA is provided in 10mM Tris-Cl, pH 8.25
3454
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
10 μg
Special Offers
For Reasearch Use Only. Not for use in Humans.