Learn More
R&D Systems™ Human IFN-beta XL Luminex™ Performance Assay
Brand:  R&D Systems™ LUXLM814
Product Code. 30001118
  • 227.00€ / 100 tests
Estimated Shipment: 20-03-2025
to see stock.
Explore more special offers
Add to basket
Specifications
Magnetic Bead-based Multiplex Assay for the Luminex™ MAGPIX™ Platform
Human
3456
1 Kit
IFN-beta
Store the unopened product at 2 - 8 °C. Do not use past expiration date.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
R&D Systems™ Human IFN-beta XL Luminex™ Performance Assay > 1 Kit