missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human IFN-gamma DuoSet ELISA

Human IFN-gamma DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

351.00€ - 685.00€

Specifications

Assay Range 9.38 - 600pg/mL
Immunoassay Kit Format 96-well Plate
Product Type Solid Phase Sandwich ELISA
Sample Type Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum
Target Species Human
View More Specs

Products 3
Product Code Brand Sufficient For Price Quantity & Availability  
Product Code Brand Sufficient For Price Quantity & Availability  
30021477
View Documents
R&D Systems™
DY285B
15 x 96-well Plates
685.00€
1 set
Please to purchase this item. Need a web account? Register with us today!
18794767
View Documents
R&D Systems™
DY285B
15 x 96-well Plates
685.00€
Pack of 15
Please to purchase this item. Need a web account? Register with us today!
30000522
View Documents
R&D Systems™
DY285B-05
5 x 96-well Plates
351.00€
Pack of 5
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The Human IFN-gamma DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human IFN-gamma.
Specifications

Specifications

9.38 - 600pg/mL
Solid Phase Sandwich ELISA
Human
96 well Micro Plates, PBS, Plate Sealers, Reagent Diluents Concentrate 2, Stop Solution, Substrate Solution, Wash Buffer
Interferon gamma
96-well Plate
Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum
3458 (Human); 15978 (Mouse); 25712 (Rat); 396991 (Porcine); 281237 (Bovine); 403801 (Canine); 493965 (Feline)
Store the unopened product at 2° to 8°C. Do not use past expiration date.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
R&D Systems™ Human IFN-gamma DuoSet ELISA

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.