missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human IL-1ra/IL-1F3 Magnetic Luminex™ Performance Assay
Magnetic bead-based multiplex assay for the Luminex™ MAGPIX™ platform
Brand: R&D Systems™ LUHM280
This item is not returnable.
View return policy
Description
- Magnetic format allows for easier wash steps - Undergoes similar validation testing as Quantikine™ ELISA Kits - Precise and Specific - Requires a small sample volume (<50μL) - Run your assay in just 3-5 hours - Mass-calibrated standards for consistent results with every new lot of material - Enough reagents for one 96-well plateSpecifications
Magnetic Bead-based Multiplex Assay for the Luminex™ MAGPIX™ Platform | |
Human | |
Magnetic | |
23.00 - 5500pg/mL | |
IL1RN | |
Cell Culture Supernates (50μL),Serum (13μL),EDTA Plasma (13μL),Heparin Plasma (13μL) | |
Store the unopened product at 2° to 8°C. Do not use past expiration date. |
1 Kit | |
IL-1ra/IL-1F3/IL1RN | |
10.91pg/mL | |
3557 | |
Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum | |
|