Learn More
Human IL-2 ELISPOT Pair, BD
Convenient set of unlabelled anti-IL-2 capture antibody and a biotinylated anti-IL-2 detection antibody
Brand:  BD Biosciences 551884
Product Code. 15816728
  • 977.00€ / Each
Estimated Shipment: 11-10-2024
to see stock.
Explore more special offers
Add to basket
Includes: Human IL-2 ELISPOT Capture Antibody (no azide/low endotoxin), Human IL-2 ELISPOT Detection Antibody (biotinylated) and a Certificate of Analysis that provides lot-specific optimal reagent concentrations