missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human IL-33 High Sensitivity Magnetic Luminex™ Performance Assay
Magnetic bead-based multiplex assay for the Luminex™ MAGPIX™ platform
Brand: R&D Systems™ LBHS3625
This item is not returnable.
View return policy
Description
- Magnetic format allows for easier wash steps
- Undergoes similar validation testing as Quantikine™ ELISA Kits
- Precise and Specific
- Requires a small sample volume (<50 μL)
- Run your assay in just 3-5 hours
- Mass-calibrated standards for consistent results with every new lot of material
- Enough reagents for one 96-well plate

Specifications
Magnetic Bead-based Multiplex Assay for the Luminex™ MAGPIX™ Platform | |
Human | |
Magnetic | |
2.44 - 10000pg/mL (Serum, Heparin Plasma, EDTA Plasma), 2.44 - 2500pg/mL (Cell Culture Supernates) | |
IL33 | |
Cell Culture Supernates (100μL),Serum (50μL),EDTA Plasma (50μL),Heparin Plasma (50μL) | |
Store the unopened product at 2° to 8°C. Do not use past expiration date. |
1 Kit | |
IL-33 | |
1.06pg/mL | |
90865 | |
Serum, Plasma (EDTA, Heparin), Cell Culture Supernatant | |
|
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
R&D Systems™ Human IL-33 High Sensitivity Magnetic Luminex™ Performance Assay > 1 Kit
Spot an opportunity for improvement?Share a Content Correction