Learn More
R&D Systems™ Human IL-7 XL Luminex™ Performance Assay
Brand:  R&D Systems™ LUXLM207
Product Code. 18705928
  • 227.00€ / 100 tests
Estimated Shipment: 17-01-2025
to see stock.
Explore more special offers
Add to basket
Specifications
Magnetic Bead-based Multiplex Assay for the Luminex™ MAGPIX™ Platform
Human
3574
1 Kit
IL-7
Store the unopened product at 2 - 8 °C. Do not use past expiration date.