Learn More
R&D Systems™ Human Kidney Biomarker Panel Luminex™ Performance Control
Luminex bead-based multiplex assays are designed to provide accurate, reproducible results for every target analyte.
Brand:  R&D Systems™ QC18
Product Code. 18412060
  • 195.00€ / 1 set
Estimated Shipment: 10-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For use as quantitative controls for the determination of human cytokine concentrations in biological fluids.
  • Concentrations have been assigned using R&D Systems Luminex™ High Performance Assays.
  • Controls are prepared in a buffered protein solution with preservatives. They contain recombinant human cytokines at low, medium and high concentrations. Controls are supplied lyophilized.
Specifications
Specifications
ELISA Kit
Human
Human Kidney Biomarker Panel
1 Ea.
Luminex Assay Controls
Store the unopened product at 2 - 8 °C. Do not use past expiration date.