Learn More
Novus Biologicals™ Human Lung Tissue MicroArray (Tumor)
Tissue arrays are 4 - 6 um in thickness, and mounted on positively charged glass slides
Brand:  Novus Biologicals™ NBP2-42077
Product Code. 18606577
  • 737.00€ / Pack of 5
Estimated Shipment: 23-12-2024
Specifications
5 Slides
Human
Immunohistochemistry, Immunohistochemistry-Paraffin