missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human MMP-7 Magnetic Luminex™ Performance Assay
Luminex bead-based multiplex assays are designed to provide accurate, reproducible results for every target analyte.
Brand: R&D Systems™ LMPM907
This item is not returnable.
View return policy
Description
- Magnetic format allows for easier wash steps
- Undergoes similar validation testing as Quantikine™ ELISA Kits
- Precise and Specific
- Requires a small sample volume (<50 μL)
- Run your assay in just 3-5 hours
- Mass-calibrated standards for consistent results with every new lot of material
- Enough reagents for one 96-well plate

Specifications
Magnetic Bead-based Multiplex Assay for the Luminex™ MAGPIX™ Platform | |
Human | |
Magnetic | |
76.60 to 55850 pg/mL | |
MMP7 | |
Cell Culture Supernates (10μL),Serum (10μL),Heparin Plasma (10μL),Platelet-poor Heparin Plasma (10μL),Saliva (10μL),Urine (10μL) | |
|
1 Kit | |
MMP-7 | |
6.6 pg/mL | |
4316 | |
Cell Culture Supernatants, Serum, Plasma (Heparin), Platelet-poor Plasma (Heparin), Saliva, Urine | |
EC 3.4.24, EC 3.4.24.23, matrilysin, Matrin, matrix metallopeptidase 7 (matrilysin, uterine), matrix metalloproteinase 7 (matrilysin, uterine), Matrix metalloproteinase-7, MMP7, MMP-7, MPSL1, PUMP1, PUMP-1, Pump-1 protease, uterine matrilysin, Uterine metalloproteinase | |
Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
R&D Systems™ Human MMP-7 Magnetic Luminex™ Performance Assay > 1 Kit
Spot an opportunity for improvement?Share a Content Correction