Learn More
R&D Systems™ Human MTF2 (NP_031384.1) VersaClone cDNA
Brand: R&D Systems™ RDC3606
This item is not returnable.
View return policy
Specifications
MTF2 | |
22823 | |
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. | |
10 μg |
NP_031384.1 | |
M96, metal response element binding transcription factor 2, PCI2, PCL2 | |
Human |