Learn More
Novus Biologicals™ Human Multi-tissue Tissue MicroArray (Normal)
The panel contains 5 to 8 different tissues on one slide.
Brand:  Novus Biologicals™ NBP2-78113
Product Code. 18653731
  • 910.00€ / Pack of 5
Estimated Shipment: 30-01-2025
Specifications
5 Slides
Human
Please bake slides at 60C for 30 minutes before use.