Learn More
Novus Biologicals™ Human Ovary Tissue MicroArray (Tumor)
Tissue arrays are 4 - 6 um in thickness, and mounted on positively charged glass slides
Brand:  Novus Biologicals™ NBP2-42067
Product Code. 18669208
  • 459.00€ / Pack of 2
Estimated Shipment: 30-01-2025
Specifications
2 Slides
Human
Immunohistochemistry, Immunohistochemistry-Paraffin