Learn More
R&D Systems™ Human PMEL (NP_008859.1) VersaClone cDNA
Contains gene inserts that are fully sequenced and produced with high quality
Brand:  R&D Systems™ RDC3162
Product Code. 18703337
  • 732.00€ / 10µg
Estimated Shipment: 28-11-2024
Specifications
PMEL17/SILV
10 μg at 0.2 μg/μL
1999 bp
D12S53EP1, gp100, ME20, ME20M, ME20-M, melanocyte protein mel 17, Melanocyte protein Pmel 17, Melanocytes lineage-specific antigen GP100, Melanoma-associated ME20 antigen, melanosomal matrix protein17, PMEL17, PMEL17P100, premelanosome proteinME20M, SI, SIL, SILV, silver (mouse homolog) like, silver homolog (mouse), Silver locus protein homolog, silver, mouse, homolog of, SILVPmel17
Human
NP_008859.1
cDNA is provided in 10 mM Tris-Cl, pH 8.5
6490
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
10 μg
Sugerencias de productos
Sugerencias de productos
Videos
SDS
Documentos
Documentos
Certificados
Ofertas especiales
Ofertas especiales