Learn More
R&D Systems™ Human Slit3 (NP_003053) VersaClone cDNA
R&D Systems™ Human Slit3 (NP_003053) VersaClone cDNA is a secreted protein and a member of the slit family of large secreted axon guidance molecules.
Brand:  R&D Systems™ RDC2420
Product Code. 18705978
  • 1170.00€ / 10µg
Estimated Shipment: 14-11-2024
Description
The Human Slit3 (NP_003053) VersaClone cDNA clone contains the complete ORF for Slit3, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The Slit3 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Specifications
Specifications
hSLIT3
10 μg at 0.2 μg/μL
4585bp
6586
Human
NP_003053
cDNA is provided in 10 mM Tris-Cl
8.5
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.