Learn More
R&D Systems™ Human UNC5H4 (NP_543148) VersaClone cDNA
R&D Systems™ Human UNC5H4 (NP_543148) VersaClone cDNA belongs to the immunoglobulin (Ig) superfamily.
Brand:  R&D Systems™ RDC2136
Product Code. 18704759
  • 747.00€ / 10µg
Estimated Shipment: 02-12-2024
Description
The Human UNC5H4 (NP_543148) VersaClone cDNA clone contains the complete ORF for UNC5H4/UNC5D, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The UNC5H4/UNC5D insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Specifications
Specifications
hUNC5D
10 μg at 0.2 μg/μL
2875bp
137970
Human
NP_543148
cDNA is provided in 10 mM Tris-Cl
8.5
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.