Learn More
R&D Systems™ Human XL Cytokine Luminex™ Performance Panel Premixed Kit
Brand: R&D Systems™ FCSTM18B-40
This item is not returnable.
View return policy
Specifications
40 Plex | |
Human | |
ELISA | |
Store the unopened product at 2°C to 8°C. Do not use past expiration date. |