Learn More
R&D Systems™ Human XL Cytokine Luminex™ Performance Panel Premixed Kit
Brand:  R&D Systems™ FCSTM18B-12
 View more versions of this product
Product Code. 30000757
  • 3570.00€ / Each
Estimated Shipment: 20-03-2025
Specifications
12 Plex
Human
ELISA
Store the unopened product at 2°C to 8°C. Do not use past expiration date.