Learn More
R&D Systems™ Human XL Cytokine Luminex™ Performance Panel Premixed Kit
Brand: R&D Systems™ FCSTM18B-05
1632.00 EUR valid until 2025-03-28
Use promo code "25306" to get your promotional price.
This item is not returnable.
View return policy
Description
Specifications
5 Plex | |
Human | |
ELISA | |
Store the unopened product at 2°C to 8°C. Do not use past expiration date. |