missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrogen Peroxide Solution, 35%, USP Grade, Chem-Lab
Click to view available options
Quantity:
1 L
Description
Hydrogen peroxide’s oxidizing properties are so strong that it is considered a highly reactive oxygen species. Due to it being a weak acid, it can form hydroperoxide or peroxide salts. Industrially, it is used for pulp/paper bleaching, in the manufacture of sodium percarbonate and sodium perborate, and in water purification to remove organic impurities and odors.
Specifications
Specifications
Color | Colorless |
CAS | 7722-84-1 |
Boiling Point | 110°C |
Melting Point | -24°C |
Molecular Formula | H2O2 |
Composition | 35+% H2O2 |
Density | 1.13g/mL |
Synonym | oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone |
Solubility Information | Water Soluble |
InChI Key | MHAJPDPJQMAIIY-UHFFFAOYSA-N |
Show More |
Safety and Handling
Product Identifier
- Hydrogen peroxide 35 weight % solution (stab.) a.r.
- Danger
- Oxidising liquid Category 1
- Acute toxicity Category 4
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 3
- H271-May cause fire or explosion; strong oxidiser.
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
- H412-Harmful to aquatic life with long lasting effects.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P310-Immediately call a POISON CENTER/doctor/.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- MIXTURE LIST-Contains : Hydrogen peroxide
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'