Learn More
HOPEN Hygiene PP Gown with Collar, Navy Blue
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
Description
- Hygiene PP Gown
- Classic with collar
- Elasticated wrists
- With one interior pocket on the left side
- With zip fastener
- 40 g/m2
- Navy Blue
Specifications
Specifications
Size | 2X-Large |
Material | Polypropylene |
Color | Navy |
Cuff Style | Elastic |
Includes | Collar |
Promotions
-
Multiple Manufacturers
Gear Up for Safety
Save up to 60% on gloves, apparel, and morePromo Code: 24131Expires: 2025-06-08
Product Content Correction
Product Title
HOPEN Hygiene PP Gown with Collar, Navy Blue >